Learn More
Medchemexpress LLC EZH2-IN-15 | 2098545-98-1 | C32H44N4O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15319110MM/1ML
Encompass_Preferred
EZH2-IN-15 is an EZH2 inhibitor that demonstrates anti-tumor activity. It is suitable for research on H3K27me3-dependent tumors. This compound is intended for research use only and is not to be used for diagnostic or therapeutic purposes. Storage conditions: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month.
  • Functions as an EZH2 inhibitor
  • Exhibits anti-tumor activity
  • Applicable for research involving H3K27me3-dependent tumors
  • For research use only
Catalog No. 50-003-72637