Learn More
Foxx Life Sciences EZLabpure™ Sterile Disposable Bottle Top Vacuum Filters with 0.45 µm PES Membrane for Prefiltration or Clarification, 1000mL, 24/CS
Supplier:  Foxx Life Sciences 1163FLS
Encompass_Preferred
The EZLabpure™ Bottle-Top Filter Funnel Only Assembly is a sterile, single-use disposable vacuum-driven lab filtration unit for high-throughput, high-capacity work. Designed in USA and made in an ISO 13485-certified cleanroom with USP Class VI materials, it delivers fast flow, low hold-up volume, and minimal protein adsorption—ideal for cell/tissue culture filtration, media preparation filters, and bioprocessing.
Features
PES membrane for fast, high-throughput filtration and low protein binding
Available pore sizes: 0.1 µm for mycoplasma removal; 0.22 µm for sterile media/reagent filtration; 0.45 µm for clarification/debris removal
Gamma sterilized (SAL 10⁻⁶), non-pyrogenic, and individually packaged
Accessory: Patented holder options including low-profile pedestal base (1116-RLS) and ring-stand cradle (1122-RLS)—both enabling hands-free vacuum filtration, leak-proof connection, and reduced contamination risk in tissue culture hoods
Catalog No. 50-001-42644