Learn More
Apexbio Technology LLC EZLink Sulfo-NHS-SS-Biotin Kit, 10 reactions
SDP
Supplier:  Apexbio Technology LLC K100610
Encompass_Preferred
The EZLink Sulfo-NHS-SS-Biotin Kit contains reagents sufficient for 10 biotin labeling reactions. It is a water-soluble biotin reagent for labeling protein, antibody or other molecules with amidogen (NH2-). The biotin in the kit is sufficient and has been activated for direct use. The whole procedure is simple and quick. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5531