Learn More
Medchemexpress LLC HY-111109 10mg , EZM 2302 CAS:1628830-21-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11110910MG
Encompass_Preferred
HY-111109 10mg EZM 2302 CAS: 1628830-21-6 EZM 2302 is an inhibitor of coactivator-associated arginine methyltransferase 1 (CARM1) with an IC50 of 6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9455