Learn More
TARGETMOL CHEMICALS INC EZM0414 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T99695MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. EZM0414 is a potent selective orally bioavailable inhibitor of SETD2 with IC50 of 18 nM in SETD2 biochemical assay and IC50 of 34 nM in a cellular assay. EZM0414 can be used for the research of relapsed or refractory multiple myeloma and diffuse large B-cell lymphoma [1]. purity: 98%
Catalog No. 50-257-8918