Learn More
Bellco Glass, Inc. EZwaste, Safety Funnel, 83mm, Stainless Steel, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800139
Encompass_Preferred
EZwaste 83B Safety Funnel, Stainless Steel features an overfill protection that will prevent spillage of liquid during filling. It comes with an attached lid that will keep VOCs from evaporating into the air. Our Safety funnel also features a removable and easy to clean flame trap that will prevent the liquid waste from igniting. Applications include but not limited Pharmaceutical, biotech, and Laboratories.Made from high quality and chemically resistant stainless steelOverfill protectionCurrently available in 83B and 120mm VersaCapSpecificationsMaterial: Stainless steelCap Size: 83BOverall Height: 16″Funnel diameter: 5.5″Funnel tube Diameter: 1.75″
Catalog No. 50-255-0623