Learn More
Bellco Glass, Inc. EZWaste Universal Stackable HPLC VersaCap 51S Solvent Waste Cap Assembly, W/ Exhaust Filter, 6 X OD Tube-3.2 mm (1/8") &1.6 mm (1/16"), 3 X Hose Barb 3.2mm(1/8") & 9.5mm(3/8"), 3 X OD Tube 12.7 mm (1/2"), 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800176
Encompass_Preferred
EZWaste Universal Stackable HPLC VersaCap 51S Solvent Waste Cap Assembly, W/ Exhaust Filter, 6 X OD Tube-3.2 mm (1/8") &1.6 mm (1/16"), 3 X Hose Barb 3.2mm(1/8") & 9.5mm(3/8"), 3 X OD Tube 12.7 mm (1/2"), 1/EA
Catalog No. 50-255-0586