Learn More
Perkin Elmer US LLC EzyFit Sample Tube - Twin - Quantity 6
Supplier:  Perkin Elmer US LLC N0777462
Encompass_Preferred
This sample tube is replacement tubing for internal standard kits. It is included in the Non HF-Resistant Internal Standard Kit (N0774068).
Catalog No. 50-210-5190