Learn More
Glass Expansion EZyLok Kit
SDP
Supplier:  Glass Expansion EL1
Encompass_Preferred
EzyLok Kit. The EzyLok Kit is an easy, safe, and effective way to connect argon supply tubes to your Glass Expansion F-Series and U-Series nebulizers. Can be used with 4mm OD nylon tubing or standard 6mm OD (1/4” OD, 1/8” ID) soft walled tubing. The EzyLok connection is made with Glass Expansion’s specially formed nebulizer side arm, which allows it to be pushed and locked securely into the popular “One Touch Fitting”, that incorporates a flexible O-Ring seal. To disconnect the nebulizer, simply push on the release button located at the top of the fitting.Replacement parts include the EzyLok Connector for 4mm Tubing, P/N QSM-4; the EzyLok 6mm Hose Adaptor, P/N 70-808-0735; and the Nebulizer Hose Clip, P/N SNP-1.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2994