Learn More
Med Vet International F/Air Canister, Remove Halogenated Gases, Compact Size, Each
Supplier:  Med Vet International 80120
Encompass_Preferred
Bickford Omnicon F/AIR is a sensible answer to anesthesia gas problems in the operating room. The F/AIR anesthesia gas filter was specifically designed to remove waste anesthesia gases such as isoflurane, halothane, enflurane, etc. from the operating room environment. Height: 6in Weight of active components: 200 grams Diameter: 3 1/4in Disposable and ready to use Compact size and no maintenance Average life is 12-15 hours.
Catalog No. 50-252-2514