Learn More
Medchemexpress LLC F092 | 2250261-59-5 | MFCD28731551 | 99.79% | 359.38 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1391711MG
Encompass_Preferred
F092 is an inhibitor of hematopoietic prostaglandin D synthase (H-PGDS) with a KD value of 0.14 nM. It can be used in the study of allergic and inflammatory responses.
  • Inhibitor of hematopoietic prostaglandin D synthase (H-PGDS)
  • Features a KD value of 0.14 nM
  • Suitable for studying allergic responses
  • Suitable for studying inflammatory responses
Catalog No. 50-003-48224