Learn More
Medchemexpress LLC HY-101510 5mg Medchemexpress, F1063-0967 CAS:613225-56-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015105MG
Encompass_Preferred
Medchemexpress, HY-101510 5mg F1063-0967 CAS:613225-56-2 F1063-0967 is a Dual-specificity phosphatase 26 ( DUSP26 ) inhibitor with an IC 50 of 11.62 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2154