Learn More
Cayman Chemical F16 Fluorescent
SDP
Supplier:  Cayman Chemical 1002210 MG
Encompass_Preferred
A potential antitumor agent; binds preferentially to mitochondrial membranes and disrupts their function; induces apoptosis in HER-1/EGFR-expressing breast carcinoma cell lines; can be used to visualize mitochondria under confocal microscopy
Catalog No. 50-288-8070