Learn More
Eurofins Discoverx PathHunter F2RL3 Activated GPCR Internalization U2OS Cell Line, 2 vials. (93-0753C3)
Supplier:  Eurofins Discoverx 930753C3
Encompass
The PathHunter GPR182 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR182 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR182 and EA-tagged β-Arrestin-2. Activation of GPR182 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1248
Add to Cart
Request a Quote