Learn More
SPECTRUM QUALITY STANDARDS FP230-L [239C Cleveland Open Cup] | Liter
Supplier:  SPECTRUM QUALITY STANDARDS FP230L
Encompass_Preferred
This standard is made for the ASTM D92 Cleveland Open Cup Flash Point with a nominal flash point of 230°C.
Catalog No. NC1716520