Learn More
Medchemexpress LLC FA-Gly-Leu-NH2 | 26400-33-9 | 99.5% | 307.34 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13730310MM/1ML
Encompass_Preferred
FA-Gly-Leu-NH2 (FAGLA) is a dipeptide substrate for proteases that can be hydrolyzed by thermophilic and neutral proteases. It shows pH-dependent variations in kcat/Km and is intended for research use only.
  • Dipeptide substrate for proteases
  • Hydrolyzed by thermophilic proteases
  • Hydrolyzed by neutral proteases
  • Shows pH-dependent variations in kcat/Km
  • Soluble in DMSO at ≥ 350 mg/mL
Catalog No. 50-003-50393