Learn More
Medchemexpress LLC FA-Gly-Leu-NH2 | 26400-33-9 | 99.5% | 307.34 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137303100MG
Encompass_Preferred
FA-Gly-Leu-NH2 (FAGLA) is a dipeptide substrate for proteases that can be hydrolyzed by thermophilic proteases and neutral proteases, showing pH-dependent variations in kcat/Km. It appears as a solid, ranging in color from white to light yellow.
  • Dipeptide substrate for proteases
  • Hydrolyzed by thermophilic proteases
  • Hydrolyzed by neutral proteases
  • Exhibits pH-dependent variations in kcat/Km
  • Soluble in DMSO at concentrations of 350 mg/mL or greater
Catalog No. 50-003-52131