Learn More
Medchemexpress LLC FA-Gly-Leu-NH2 (FAGLA) | 26400-33-9 | 99.5% | 307.34 | 25 MG

Supplier: Medchemexpress LLC 25MGHY137303
FA-Gly-Leu-NH2 (FAGLA) is a dipeptide substrate for proteases that can be hydrolyzed by thermophilic proteases and neutral proteases, showing pH-dependent variations in kcat/Km.
- Formula: C15H21N3O4
- Appearance: Solid
- Color: White to light yellow
- Soluble in DMSO: ≥ 350 mg/mL
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.