Learn More
Medchemexpress LLC FA-Leu-Gly-Pro-Ala-OH | 78832-65-2 | 99.4% | 476.52 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP26615MG
Encompass_Preferred
FA-Leu-Gly-Pro-Ala-OH is a furylacryloyl-terminal tetrapeptide that serves as a substrate for bacterial collagenase and spirochete metalloendopeptidase. It is specifically hydrolyzed by spirochete collagenase only at the Leu-Gly bond. This compound can be used to determine the equilibrium constant of peptide bond hydrolysis and to detect collagenase-mediated cleavage reactions via turbidimetry based on absorbance reduction.
  • Substrate for bacterial collagenase and spirochete metalloendopeptidase.
  • Specifically hydrolyzed at the Leu-Gly bond by spirochete collagenase.
  • Useful for determining the equilibrium constant of peptide bond hydrolysis.
  • Can detect collagenase-mediated cleavage reactions via turbidimetry.
Catalog No. 50-004-00383