Learn More
Medchemexpress LLC HY-79511 500mg Medchemexpress, FAAH-IN-2 CAS:184475-71-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY79511500MG
Encompass_Preferred
Medchemexpress, HY-79511 500mg FAAH-IN-2 CAS:184475-71-6 FAAH-IN-2 (O-Desmorpholinopropyl Gefitinib) is a potent FAAH(fatty acid amide hydrolase) inhibitor extracted from Patent WO/2008/100977A2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2296