Learn More
ACROBiosystems Recombinant Protein;Human FABP1 / L-FABP Protein;E.coli;200UG;FA1,FABP1,FABPL,L-FABP,FABP-1,FABP-L
Supplier:  ACROBiosystems FA1H5148200UG
Encompass_Preferred
Recombinant Protein;Human FABP1 / L-FABP Protein;200UG;Human FABP1, His Tag (FA1-H5148) is expressed from E.coli cells. It contains AA Ser 2 - Ile 127 (Accession # NP_001434).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8256