Learn More
ABclonal Technology FABP5 Rabbit mAb
SDP
Supplier:  ABclonal Technology A27255100UL
Encompass_Preferred
This gene encodes the fatty acid binding protein found in epidermal cells and was first identified as being upregulated in psoriasis tissue Fatty acid binding proteins are a family of small highly conserved cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands FABPs may play roles in fatty acid uptake transport and metabolism Polymorphisms in this gene are associated with type 2 diabetes The human genome contains many pseudogenes similar to this locus
Catalog No. 50-000-11520