Learn More
Aviva Systems Biology Corp FABP7 Antibody - N-terminal region (ARP33827_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33827_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FABP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FABP7. Alternate names: MRG, BLBP, FABPB, B-FABPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55163