Learn More
Medchemexpress LLC FABPs ligand 6 | 2988135-14-2 | 99.9% | C28H27FN2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY145990100MG
Encompass_Preferred
FABPs ligand 6 (MF6) is an FABP5 and FABP7 inhibitor with KD values of 874 nM for FABP5 and 20 nM for FABP7. This compound rescues mitochondrial function by blocking voltage-dependent anion channel (VDAC)-1-dependent mitochondrial macropore formation, a mechanism involved in FABP5-mediated injury. It can be used for multiple sclerosis research, improving EAE severity and attenuating oxidative levels and inflammatory responses in mouse models.
  • FABP5 and FABP7 inhibitor
  • Rescues mitochondrial function
  • Attenuates oxidative levels
  • Inhibits inflammatory responses
  • Protects oligodendrocytes
  • Suitable for multiple sclerosis research
Catalog No. 50-003-55309