Learn More
Medchemexpress LLC FABPs ligand 6 (MF6) | 2988135-14-2 | 99.9% | C28H27FN2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14599050MG
Encompass_Preferred
FABPs ligand 6 (MF6) is an inhibitor of FABP5 and FABP7, with KD values of 874 nM and 20 nM, respectively. It is utilized in multiple sclerosis research, where it can restore mitochondrial function by inhibiting voltage-dependent anion channel (VDAC)-1-dependent mitochondrial macropore formation. In vivo studies have shown that MF6 can improve the severity of experimental autoimmune encephalomyelitis (EAE) and reduce oxidative levels and inflammatory responses.
  • Inhibits FABP5 and FABP7
  • Rescues mitochondrial function
  • Reduces oxidative levels
  • Attenuates inflammatory response
  • Used in multiple sclerosis research
Catalog No. 50-003-55268