Learn More
Fisherbrand™ Face Masks
Disposable face masks feature a thin middle layer of meltblown filter media with spunbonded polypropylene inner and outer layers.
Description
- Bacterial filtration efficiency is 99%
- Layers ultrasonically welded to reduce particle generation
- Blue outer layer with white inner layer
- Elastic ear loops