Learn More
Grainger Face Mill MFPN 452500R6T MFPN 2.5000
Supplier:  Grainger 170KC2
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter designed for high-speed machining of steel, cast iron, and non-ferrous metals. It features a 2-inch diameter with 4 carbide inserts for a maximum depth of cut of 1 inch. The cutter has a 45 degree lead angle and is balanced for speeds up to 15,000 RPM. The carbide inserts utilize a positive rake angle for smooth, efficient cutting and feature TiAlN nano-grain PVD coating for heat and wear resistance. This versatile face mill is well-suited for facing, slotting, and contouring operations. With its rigid design and sharp cutting edges, the Kyocera Face Mill delivers accurate, repeatable milling performance.
Catalog No. 50-276-4247