Learn More
Grainger Face Mill MFPN 454000R6T MFPN 400
Supplier:  Grainger 170KA7
Encompass_Preferred
The Kyocera Face Mill is a high-performance face milling cutter with a diameter of 2 inches. It features 4 indexable carbide inserts with a TiAlN PVD coating for wear resistance and heat resistance. The cutter body is made of steel and has a Weldon flat shank with a 7/8 inch shank diameter. It is capable of face milling a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The maximum depth of cut is 0.630 inches and it has a cutting speed range of 65-325 SFM. This durable and versatile face mill is well-suited for heavy roughing and finishing operations.
Catalog No. 50-276-4276