Learn More
Medchemexpress LLC Factor B-IN-1 | 1481631-75-7 | 95.0% | 332.36 g·mol⁻¹ | C19H16N4O2 | 5 MG

Supplier: Medchemexpress LLC HY1365565MG
Factor B-IN-1 is a research-grade small-molecule inhibitor of complement Factor B intended for in vitro and in vivo studies. The compound has formula C19H16N4O2, molecular weight 332.36 g·mol⁻1, CAS 1481631-75-7, and is supplied at high purity for laboratory use. Refer to the product datasheet for handling, solubility, and storage instructions.
- Small-molecule inhibitor of complement Factor B.
- Molecular formula C19H16N4O2; molecular weight 332.36 g·mol⁻1.
- Purity typically 95.0%.
- Soluble in DMSO (≈50 mg/mL) and formulatable for in vivo use.
- Powder storage at -20°C; store solutions at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.