Learn More
Genetex Inc Factor XIIIa Monoclonal Antibody [MSVA-813R] HistoMAX™ (GTX04452) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04452
Encompass_Preferred
The Factor XIIIa antibody [MSVA-813R] HistoMAX™ (GTX04452) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human lymph node tissue using GTX04452 Factor XIIIa antibody [MSVA-813R] HistoMAX. Strong factor XIII immunostaining of sinus histiocytes in a lymph node.
Catalog No. NC3667725