Learn More
ATCC Faecalibacterium prausnitzii
Supplier:  ATCC 27766
Encompass
Faecalibacterium prausnitzii strain VPI C13-20-A was isolated from human feces. This strain is whole-genome sequenced.
Catalog No. 50-238-2954