Learn More
MSC Fairbanks Rigid Top Plate Caster, 6 in Wheel, Polyurethane, 1000 lb Capacity
Supplier: MSC 01357615
This rigid top plate caster features a 6-inch diameter wheel made of polyurethane with a semi-steel center. It is designed for institutional and industrial applications, offering a substantial load capacity of 1000 lb. Its overall height is 7-1/4 inches, making it suitable for various demanding environments.
- Rigid top plate mount
- 6 in wheel diameter
- 2 in wheel width
- Polyurethane wheel material with semi-steel center
- 1000 lb load capacity
- 7-1/4 in overall height
- Designed for institutional and industrial applications
- Orbit formed bolt