Learn More
Medchemexpress LLC Fak-in-14 | 2766666-22-0 | 99.8% | 502.43 g·mol⁻¹ | C21H24BrN7OS | 10 MG

Supplier: Medchemexpress LLC HY15543910MG
FAK-IN-14 is a small-molecule inhibitor of focal adhesion kinase (FAK) with a reported biochemical potency of IC50 0.2438 nM. It has been reported to induce early apoptosis in U87-MG cells and to arrest the cell cycle at the G2/M phase. The compound is supplied as a white to off-white solid for in vitro research and is highly pure.
- Potent FAK inhibition (IC50 0.2438 nM).
- Induces early apoptosis in U87-MG cells and causes G2/M cell cycle arrest.
- High purity (99.81%) and white to off-white solid appearance.
- Soluble in DMSO at 25 mg/mL; ultrasonic assistance may be required.
- Recommended storage: powder at -20°C for long-term stability.
- Intended for in vitro research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.