Learn More
Abcam FAK Inhibitor 14, FAK autophosphorylation inhibitor, 50MG
Supplier:  Abcam AB14450350MG
Encompass_Preferred
MW 174.63 Da. Selective focal adhesion kinase (FAK) autophosphorylation inhibitor (IC₅₀ = 1 μM). No significant activity at other kinases including EGFR, PDGFR and IGF-RI. Increases cell detachment and inhibits cell adhesion in cancer cells. Anticancer agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5664