Learn More
Medchemexpress LLC FAK ligand-Linker Conjugate 1 | 2307461-45-4 | 98.9% | 583.58 | 5 MG

Supplier: Medchemexpress LLC HY441485MG
FAK ligand-Linker Conjugate 1 incorporates a ligand for FAK and a PROTAC linker, which recruits E3 ligases (such as VHL, CRBN, MDM2, and IAP). It can be extensively used for PROTAC-mediated protein degradation.
- Purity: 98.91%
- Appearance: Solid
- Color: White to off-white
- Solubility: In Vitro: DMSO : 100 mg/mL (171.36 mM; Needs ultrasonic; Hygroscopic DMSO has a significant impact on the solubility of the product, so newly opened DMSO should be used).
- Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 2 years, -20°C for 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.