Learn More
Medchemexpress LLC Fak1 protein, human (Sf9, GST) | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70168250UG
Encompass_Preferred
Recombinant full-length human focal adhesion kinase 1 expressed in Sf9 insect cells and supplied as an N-terminal GST-tagged, purified protein in a buffered solution. Activity is verified by ADP-Glo assay; the product is formulated for frozen storage and research use only.
  • Full-length human sequence (M1-H1052).
  • N-terminal GST tag for affinity purification and detection.
  • Expressed in Sf9 insect cells to support eukaryotic folding.
  • Activity verified by ADP-Glo biochemical assay.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Supplied in HEPES buffer with 20% glycerol and 1 mM DTT for stability.
Catalog No. 50-001-75567