Learn More
Medchemexpress LLC Falnidamol | 196612-93-8 | 99.4% | 387.84 g/mol | C18H19ClFN7 | 1 ML

Supplier: Medchemexpress LLC HY1032210MM 1ML
Falnidamol (BIBX 1382) is a small-molecule EGFR tyrosine kinase inhibitor supplied for research use. It is provided in both solid and solution formats for biochemical and cellular assays; the solution form is commonly offered as 10 mM in DMSO (1 mL).
- Selective EGFR inhibitor with IC50 ≈ 3 nM.
- Also shows ErbB2 activity (IC50 ≈ 3.4 μM).
- Chemical formula C18H19ClFN7 and molecular weight 387.84 g/mol.
- Available as 10 mM solution in DMSO (1 mL) and as solids in multiple quantities.
- Purity ~99.4% by HPLC.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.