Learn More
Medchemexpress LLC Falnidamol | 196612-93-8 | MFCD09838900 | 99.4% | 387.84 g·mol⁻¹ | C18H19ClFN7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1032250MG
Encompass_Preferred
Falnidamol (BIBX 1382) is an orally active, selective epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor used in preclinical research to probe EGFR-dependent signaling and cancer cell proliferation. It exhibits potent EGFR inhibition and is supplied as a characterized research compound for laboratory studies.
  • Potent EGFR inhibition with low nanomolar activity.
  • Useful for studies of EGFR-driven cancer cell signaling.
  • Supplied as a high-purity, analytically characterized research chemical.
  • Available in small quantities suitable for screening and dosing experiments.
Catalog No. 50-002-45958