Learn More
Medchemexpress LLC HY-B1878 1g Medchemexpress, Faltan CAS:133-07-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB18781G
Encompass_Preferred
Medchemexpress, HY-B1878 1g Faltan CAS:133-07-3 Faltan is a dicarboximide fungicide , widely used on vines and several vegetable crops, and is also cytotoxic effect on human bronchial epithelial cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3025