Learn More
Medchemexpress LLC Faltan-d4 | 1327204-12-5 | 99% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1878S1MG
Encompass_Preferred
Faltan-d4 is the deuterium-labeled version of Faltan. Faltan is a dicarboximide fungicide extensively used on grapevines and various vegetable crops, and it also exhibits a cytotoxic effect on human bronchial epithelial cells. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitative analysis during drug development. Deuteration is gaining interest due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09791