Learn More
Cayman Chemical Falurdn 10mg
SDP
Supplier:  Cayman Chemical 1586710 MG
Encompass_Preferred
A nucleoside analog with antiviral activity, inhibiting thymidine kinases from herpes simplex virus types 1 and 2 with Ki values of 0.14 and 0.95 µM, while blocking green monkey Vero cell thymidine kinase less effectively (Ki = 53 µM)
Catalog No. 50-297-1593