Learn More
Aviva Systems Biology Corp FAM120C Antibody - N-terminal region (ARP60764_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60764_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM120C. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FAM120C. Alternate names: CXorf17, FLJ20506, FLJ43676, ORF34ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-152