Learn More
Aviva Systems Biology Corp FAM79B Antibody - C-terminal region (ARP55878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM79B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FAM79B. Alternate names: FAM79B, FLJ41238, FLJ43694, MGC126599, MGC126601ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-091