Learn More
Medchemexpress LLC Famciclovir | 104227-87-4 | 321.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17426100MG
Encompass_Preferred
Famciclovir (BRL 42810) is an orally active nucleoside analogue. It is an antiviral agent with potent activities against HBV, HSV, and VZV. Famciclovir can be used for the research of herpesvirus infection. A bulleted list of features and benefits was not found on the page.
Catalog No. 50-003-86586