Learn More
Medchemexpress LLC Famciclovir | 104227-87-4 | 99.69% | 321.33 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1742610MM/1ML
Encompass_Preferred
Famciclovir is an orally active nucleoside analogue and an antiviral agent. It shows potent activities against HBV, HSV, and VZV, making it suitable for research on herpesvirus infection.
  • Orally active nucleoside analogue
  • Antiviral agent
  • Potent against HBV
  • Potent against HSV
  • Potent against VZV
  • Suitable for herpesvirus infection research
Catalog No. 50-003-86616