Learn More
Med Vet International Famciclovir Tablets (Famvir), 500mg, 30ct
Supplier:  Med Vet International RXFAMCIC50030
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Famciclovir is a generic version of Famvir. It is used to treat infections caused by certain types of viruses and should be stored at room temperature away from light and moisture.
Catalog No. 50-282-7881