Learn More
Medchemexpress LLC HY-B0377 500mg Medchemexpress, Famotidine CAS:76824-35-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0377500MG
Encompass_Preferred
Medchemexpress, HY-B0377 500mg Famotidine CAS:76824-35-6 Famotidine (MK-208) is a competitive histamine H2-receptor antagonist. Its main pharmacodynamic effect is the inhibition of gastric secretion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2501