Learn More
Sigma Aldrich Fine Chemicals Biosciences famotidine United States P25MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 1269200125MG
Encompass
famotidine is a hydrophilic cationic histamine H2 receptor antagonist drug that effectively inhibits gastric acid secretion in humans. This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. for further information and support please go to the website of the issuing Pharmacopoeia.
Catalog No. 50-290-3201