Learn More
Aviva Systems Biology Corp FANCL Antibody - middle region (ARP56321_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56321_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FANCL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FANCL. Alternate names: FAAP43, FLJ10335, PHF9, POGApplicationIHC, WB. Species reactivity: Dog, Human, Rabbit
Catalog No. 50-014-61809