Learn More
Selleck Chemical LLC (+)-Fangchinoline 5mg 436-77-1 (+)-Limacine, Tetrandrine B, Hanfangichin B
SDP
Supplier:  Selleck Chemical LLC S36115MG
Encompass_Preferred
Fangchinoline ((+)-Limacine, Tetrandrine B, Hanfangichin B) is a phytochemical that has been shown to elicit anti-cancer effects in prostate and breast cancer cell lines via inducing G1 cell cycle arrest. It has also been shown to possess neuroprotective activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8426